Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD275 (B7-H2) Mouse anti-Human, Clone: GM13C1, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA117756

Catalog No. 01-670-169


Add to cart

Description

Description

This antibody recognizes native human ICOSL protein expressed in transfected mammalian cells.

Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
Specifications

Specifications

CD275 (B7-H2)
Monoclonal
1 mg/mL
PBS with 0.01% sodium azide; pH 7.2
O75144
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, KIAA0653, LICOS, ICOS Ligand
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23308
ELISA, Flow Cytometry
GM13C1
Unconjugated
ICOSLG
Liquid
ICOSLG
cDNA encoding human ICOSL
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit