Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD28, Clone: CD28.2; Beckman Coulter™

$461.04

Specifications

Antigen CD28
Clone CD28.2
Format Purified
Host Species Murine
Isotype IgG1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
COIM1376
View Documents
Coulter Corporation
IM1376
0.2mg Each for $461.04
Add to cart
 
Description

Description

The CD28 antigen (Tp44) is a homodimer of disulfide-linked chains, with a molecular weight of 44 kDa each. CD28 antigen is expressed by a majority of CD3+ T lymphocytes. CD4+ T lymphocytes express the CD28 antigen more frequently than do CD8+ T lymphocytes. CD28 antigen is also present on plasma cells and thymocytes. This antigen is involved in the adhesion of T lymphocytes with B lymphocytes, through the B7/BB-1 (CD80) or B70/B7-2 (CD86) glycoproteins and provides a major co-stimulatory signal for T cell activation.

Host Species: Mouse
Clone: CD28.2
Isotype: IgG1
Cross Reactivity: Human

Flow Cytometry

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

CD28
Purified
IgG1
Primary
CD28.2
Murine
Monoclonal
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit