Learn More
CD29 Mouse anti-Human, Brilliant Violet 786, Clone: MAR4., BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 564815
Catalog No. BDB564815
Add to cart
Description
Description
The MAR4 monoclonal antibody specifically binds to CD29. CD29 is a 130 kDa integrin β 1 subunit that is expressed as a heterodimeric complex with one of six distinct α subunits, comprising the very late activation antigen (VLA) subfamily of adhesion receptors. The β 1 subunit has a broad tissue distribution; it is expressed on lymphocytes, monocytes and weakly on granulocytes, but not on erythrocytes. These receptors are involved in a variety of cell-cell and cell-matrix interactions.
Specifications
Specifications
CD29
MAR4.
ITGB1; Integrin beta-1; Integrin β1; FNRB; GPIIA; ITB1; MDF2; MSK12; VLA-4β
Affinity purified
Purified Human α5β1 Fibronectin Receptor
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide
Murine
IgG1, κ
RUO
Primary
Human