Learn More
CD3e Mouse anti-Human, Biotin, Clone: M2AB, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112008
Catalog No. 01-670-731
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation.
Specifications
Specifications
CD3e
Monoclonal
Biotin
CD3E
Liquid
CD3E
human thymocytes
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
M2AB
PBS with 0.2% carrier protein and 0.08% sodium azide
P07766
FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Mouse
IgG1
100 Tests
4° C, do not freeze
Primary
916