Learn More
CD3/CD19/CD45 Mouse anti-Human, FITC, PE, PE-Cyanine5, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112481
Catalog No. 01-674-550
Add to cart
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1), PE-conjugated CD19 (clone 1G9, isotype IgG1) and Cy5-PE-conjugated CD45 (clone BHPT-1, isotype IgG1).
T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. CD19 assembles with the antigen receptor of B-lymphocytes in order to decrease the threshold for antigen receptor-dependent stimulation. CD45 acts as a positive regulator of T-cell coactivation upon binding to DPP4. The first PTPase domain has enzymatic activity, while the second one seems to affect the substrate specificity of the first one. Upon T-cell activation, CD45 recruits and dephosphorylates SKAP1 and FYN.
Specifications
Specifications
CD3/CD19/CD45
Cocktail
FITC, PE, PE-Cyanine5
CD3E
Liquid
CD19, CD3D, CD3E, CD3G, PTPRC
human thymocytes and human peripheral blood monocytes from a rheumatoid arthritis patient
Affinity chromatography
RUO
Antibody
5788, 915, 916, 917, 930
Flow Cytometry
1G9
PBS with 0.2% carrier protein and 0.1% sodium azide
P04234, P07766, P08575, P09693, P15391
CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, AW495831, B-lymphocyte antigen CD19, differentiation antigen CD19, PTPRC, B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Mouse
IgG1
50 Tests
4° C, do not freeze
Primary
Human