Learn More
CD3/CD25 Mouse anti-Human, FITC, PE, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112091
Catalog No. 01-670-810
Add to cart
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1) and PE-conjugated CD25 (clone ITYV, isotype IgG1).
T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells. To date, a total of 151 CD clusters and subclusters have been identified.
Specifications
Specifications
CD3/CD25
Cocktail
FITC, PE
CD3D
Liquid
CD3D, CD3E, CD3G, IL2RA
human thymocytes and PHA-activated human T-lymphocytes
Protein A/G
RUO
Antibody
3559, 915, 916, 917
Flow Cytometry
ITYV
PBS with 0.2% carrier protein and 0.08% sodium azide
P01589, P04234, P07766, P09693
CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, IL-2R alpha, IL2 RA antibody, Il2ra antibody, Interleukin 2 receptor alpha chain antibody, TAC antigen antibody, IL-2R alpha chain
Mouse
IgG1
100 Tests
4° C, do not freeze
Primary
Human