Learn More
CD3/CD4/CD45 Mouse anti-Human, FITC, PE, PE-Cyanine5, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112469
Catalog No. 01-674-525
Add to cart
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1), PE-conjugated CD4 (clone 7E14, isotype IgG1) and Cy5-PE-conjugated CD45 (clone BHPT-1, isotype IgG1).
T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation.. The CD4 antigen is involved in the recognition of the type II MHC antigen. It is also a receptor for HIV. It is present on most T helper cells and normal thymocytes. The cytoplasmic portion of CD4 is associated with p56lck tyrosine kinase. CD4 expression is commonly found in human lymph nodes and tonsils.. CD45 Leukocyte Common Antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and maligt, non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals.
Specifications
Specifications
CD3/CD4/CD45
Cocktail
FITC, PE, PE-Cyanine5
CD3D
Liquid
CD3D, CD3E, CD3G, CD4, PTPRC
human thymocytes, human peripheral blood T lymphocytes and peripheral blood monocytes from a rheumatoid arthritis patient
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
7E14
PBS with 0.2% carrier protein and 0.08% sodium azide
P01730, P04234, P07766, P08575, P09693
CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4, PTPRC, B220, CD45, CD45R, GP180, L-CA, LCA, LY5, T200, CD45 antigen, T200 glycoprotein, T200 leukocyte common antigen, protein tyrosine phosphatase, receptor type, c polypeptide, receptor-type tyrosine-protein phosphatase C
Mouse
IgG1
100 Tests
4° C, do not freeze
Primary
5788, 915, 916, 917, 920