Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3/CD4/CD8, PE, PE-Cyanine5, clone: M2AB, 7E14, 17D8, Invitrogen™

Antibody Cocktail

Manufacturer:  Invitrogen MA112473

Catalog No. 01-674-532

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation.. The CD4 antigen is involved in the recognition of the type II MHC antigen. It is also a receptor for HIV. It is present on most T helper cells and normal thymocytes. The cytoplasmic portion of CD4 is associated with p56lck tyrosine kinase. CD4 expression is commonly found in human lymph nodes and tonsils.. CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells.

This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1), PE-conjugated CD4 (clone 7E14, isotype IgG1) and Cy5-PE-conjugated CD8 (clone 17D8, isotype IgG1).
Specifications

Specifications

CD3/CD4/CD8
Cocktail
PE, PE-Cyanine5
CD8A
Purified
CD3D, CD3E, CD3G, CD4, CD8A, CD8B
human thymocytes and human peripheral blood T lymphocytes
Affinity chromatography, Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
17D8
PBS with 0.2% carrier protein and 0.08% sodium azide
P01730, P01732, P04234, P07766, P09693, P10966
CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4, CD8, Leu-2, MAL, p32, CD8a, CD8b, CD8alpha, CD8beta
Murine
IgG1
100 Tests
Store at 4°C
Primary
915, 915, 916, 916, 917, 917, 920, 920, 925, 925, 926, 926
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit