Learn More
CD3Ε Mouse anti-Human, Mouse, Clone: APA1/1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05785
Catalog No. 05-785-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect CD3Ε using this Anti-CD3Ε Antibody, clone APA1/1 validated for use in flow cytometry, immunoprecipitation, western blotting and immunocytochemistry.
Specifications
Specifications
CD3ε
APA1/1
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide
Purified
Murine
IgG1
200μg
Primary
Human, Murine
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
Unconjugated
NM_000733
CD3E, T3E, TCRE
Purified human CD3 epsilon
Protein G purfied
2-8°C for one year
Monoclonal