Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3e Monoclonal Antibody (M2AB), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA112006
Description
CD3e Monoclonal antibody specifically detects CD3e in Human samples. It is validated for Flow Cytometry.Specifications
CD3e | |
Monoclonal | |
Unconjugated | |
CD3g | |
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; Cd247; CD247 antigen; CD247 antigen, zeta subunit; Cd247 molecule; CD3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 glycoprotein precursor; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 TCR complex; CD3 type I transmembrane glycoprotein; CD3 type I transmembrane glycoprotein precursor; CD3 zeta chain; Cd3d; CD3D antigen d | |
Mouse | |
Protein A/G | |
RUO | |
915, 916, 917, 919 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry | |
M2AB | |
PBS with 0.08% sodium azide | |
P04234, P07766, P09693, P20963 | |
Cd247, Cd3d, CD3E, CD3g | |
human thymocytes | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction