Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3, Mouse anti-Human, Clone: SK7, BV650, BD
Supplier: BD Biosciences 563999
Description
Host Species: Mouse
Clone: SK7
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Human Thymocytes
Flow cytometry
Specifications
CD3 | |
SK7 | |
Human Thymocytes | |
100 Tests | |
Primary |
Monoclonal | |
Mouse | |
Affinity Purified | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction