Learn More
anti-CD3 epsilon, Alexa Fluor 700, Clone: UCHT1, R&D Systems™
Manufacturer:  R&D Systems FAB100N025
 View more versions of this product
Catalog No. FAB100N025
Add to cart
Specifications
Specifications
CD3 epsilon
UCHT1
Flow Cytometry 5 uL/10^6 cells
Purified
CD3E
Human thymocytes (1),
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
916
Human
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD3e, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Mouse
IgG1
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Recognizes the epsilon-chain of the CD3/T cell antigen receptor complex (McMichael, A.J. et al. (1987) Leucocyte Typing III: White Cell Differentiation Antigens, Oxford University Press, New York; Knapp, W. et al. (1989) Leucocyte Typing IV: White Cell Differentiation Antigens, Oxford University Press, New York; Schlossman, S. et al. (1995) Leucocyte Typing V: White Cell Differentiation Antigens, Oxford University Press, New York).