Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3/HLA-DR, FITC, PE, clone: M2AB, 423L, Invitrogen™

Antibody Cocktail

Manufacturer:  Invitrogen MA112115

Catalog No. 01-670-834

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules. This association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation.

This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1) and PE-conjugated HLADR (clone 423L, isotype IgG2a).
Specifications

Specifications

CD3/HLA-DR
Cocktail
FITC, PE
CD3E
Purified
CD3E, CD74
human thymocytes
Protein A/G, Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
423L
PBS with 0.2% carrier protein and 0.08% sodium azide
P04233, P07766
CD3e, CD3 epsilon, FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, CD74 antigen, HLA class II histocompatibility antigen gamma chain, HLA-DR antigens-associated invariant chain, HLA-DR-gamma, Ia-associated invariant chain, MHC HLA-DR gamma chain, gamma chain of class II antigens, ia antigen-associated invariant chain, HLADG, DHLAG, IA-gamma
Murine
IgG1/IgG2a
50 Tests
Store at 4°C
Primary
916, 916, 972, 972
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit