Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300b Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525127

Catalog No. PIPA525127


Add to cart

Description

Description

CD300LB is a nonclassical activating receptor of the immunoglobulin (Ig) superfamily expressed on myeloid cells.
Specifications

Specifications

CD300b
Polyclonal
PBS with 0.09% sodium azide
A8K4G0
CMRF35-A2, CD300b, CLM-7, CLM7, CMRF35-A2, IREM-3, IREM3, TREM-5, TREM5, CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule like family member b, CMRF35-like molecule 7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
124599
Western Blot
Unconjugated
CD300LB
Liquid
CD300LB
KLH conjugated synthetic peptide between 57-85 amino acids from the N-terminal region of human CD300LB
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit