Learn More
CLEC4C Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSD00005W
Catalog No. PIOSD00005W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Specifications
Specifications
CLEC4C
Polyclonal
whole serum with no preservative
Q8WTT0
C-type lectin superfamily member 7, Blood dendritic cell antigen 2 protein, BDCA-2, Dendritic lectin, CD303 antigen, CLEC4C, BDCA2, CLECSF11, CLECSF7, DLEC, HECL, C-type lectin domain family 4 member C
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
CLEC4C
Lyophilized
CLEC4C
A synthetic peptide from extracellular domain of human CD303
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
170482