Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse anti-Human, APC-Cy7, Clone: WM59, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563653

Catalog No. BDB563653


Add to cart

Description

Description

The WM59 monoclonal antibody specifically binds to platelet endothelial cell adhesion molecule-1, (PECAM-1, PECAM1), which is also known as GPIIA', or EndoCAM. CD31 is a 130 kDa type I transmembrane glycoprotein that belongs to the Ig gene superfamily. CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, NK cells, T cell subsets, and in high amounts on endothelial cells. CD31 functions as a vascular endothelial cell adhesion molecule and is involved in the transendothelial migration of leucocytes in inflammatory responses. It may also be involved in thrombosis, angiogenesis, and wound healing.

Specifications

Specifications

CD31
WM59
EndoCAM; GPIIA; PECA1; PECAM1; Platelet endothelial cell adhesion molecule
Affinity Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
APC-Cyanine7
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Murine
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit