Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD31; Clone: MEC 13.3; BD

Manufacturer:  BD Biosciences 562861

Catalog No. BDB562861


Add to cart

Description

Description

Host Species: Rat
Clone MEC 13.3
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen 129/Sv mouse-derived endothelioma cell line tEnd.1

Flow Cytometry

Specifications

Specifications

CD31
0.2mg/mL
Affinity Purified
129/Sv mouse-derived endothelioma cell line tEnd.1
100μg
Primary
Murine
MEC 13.3
PerCP-Cyanine5.5
Rat
IgG2a
RUO
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit