Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD31, Clone: MEC 7.46, Abcam™ Non-distribution product as customer accommodation.

Manufacturer:  Abcam AB738850UG

Catalog No. NC0092411


Add to cart

Description

Description

Host Species: Rat
Clone: MEC 7.46
Isotype: IgG1, κ
Cross Reactivity: Mouse
Immunogen: Mouse t.end.1 cells (polyoma middle T (PmT)-transformed EC)

Applications:

Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation

Specifications

Specifications

CD31
0.1mg/mL
Rat
IgG1, κ
Cancer and Tumor Biology, Cardiovascular, Cell Biology, Signal Transduction, Stem Cell Biology
Monoclonal
MEC 7.46
Unconjugated
Mouse t.end.1 cells (polyoma middle T (PmT)-transformed EC)
50μg
Primary
Murine
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit