Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse anti-Human, PE-CF594, Clone: WM59, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563652

Catalog No. BDB563652


Add to cart

Description

Description

The WM59 monoclonal antibody specifically binds to a 130 kDa glycoprotein, gpIIa', also known as platelet endothelial cell adhesion molecule-1, (PECAM-1). CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, and in high amounts on endothelial cells. This molecule has been implicated in a number of cellular phenomena, including vascular wound healing and angiogenesis, and transendothelial migration of leukocytes in inflammatory responses.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

Specifications

Specifications

CD31
WM59
EndoCAM; GPIIA'; PECA1; PECAM1; Platelet endothelial cell adhesion molecule
Affinity Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit