Learn More
anti-CD31/PECAM-1, Alexa Fluor 488, Polyclonal, R&D Systems™
Manufacturer:  R&D Systems FAB3628G025
 View more versions of this product
Catalog No. FAB3628G025
Add to cart
Specifications
Specifications
CD31/PECAM-1
Alexa Fluor 488
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
PECAM1
Mouse myeloma cell line NS0-derived recombinant mouse CD31/PECAM-1, Glu18-Lys590, Accession # Q08481
Antigen Affinity-purified
RUO
Primary
5175
Mouse, Rat
Flow Cytometry
Flow Cytometry 5 uL/10^6 cells
Q08481
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule
Goat
IgG
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Polyclonal
Detects mouse CD31/PECAM-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD31 and recombinant porcine CD31 is observed. Detects mouse CD31 and rat CD31 in flow cytometry.