Learn More
CD321 (JAM-1) Rat anti-Mouse, PE, Clone: H202-106, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 564908
Description
The H202-106 monoclonal antibody specifically binds to CD321 which is also known as JAM-1 (Junctional adhesion molecule 1), Junctional adhesion molecule A (JAM-A), and F11 Receptor (F11R). CD321 is a type I transmembrane glycoprotein that includes two extracellular immunoglobulin-like domains. CD321 functions as an adhesion receptor molecule on platelets. It also supports the tight junction formation between endothelial cells, where it may regulate the transendothelial migration of leucocytes, and some epithelial cells. CD321 expression by certain subsets of tissue-derived, MHC Class II-positive thymic epithelial cells, macrophages, and dendritic cells suggest that it may play a role in the organization and function of antigen presenting cells.
Specifications
CD321 | |
Monoclonal | |
0.2mg/mL | |
F11r; F11 receptor; JAM; JAM-A; Jam1; Jcam; Jcam1; Ly106 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
H202-106 | |
PE | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Thymic Stromal Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.