Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD326 Rat anti-Mouse, APC, Clone: G8.8, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 563478

Catalog No. BDB563478


Add to cart

Description

Description

The G8.8 antibody reacts with CD326/Ep-CAM (Epithelial Cell AMolecule), also known as gp40 in the mouse and by a variety of names (including GA733-2, CO17-1A, and EGP) in the human. In the mouse, Ep-CAM is a 40-42 kDa cell-surface type 1 transmembrane glycoprotein expressed on thymic epithelial cells, thymic dendritic cells, immature thymocytes, a small subset of peripheral T lymphocytes, intestinal epithelium, kidney-collecting tubule epithelium, keratinocytes, Langerhans cells and lymph node and splenic dendritic cells. Profiles of Ep-CAM expression on fetal thymocytes and on the CD4[-] CD8[-], CD4[+] CD8[+], CD4[-] CD8[+], and CD4[+] CD8[-] subsets of adult thymocytes have been published. In unrelated studies, mouse Ep-CAM mRNA was detected in tissues containing epithelial cells (kidney, stomach, intestine, lung, and thymus) and in plasma cells and plasmacytomas, but not in heart, muscle, liver, brain, spleen, B lymphomas, or pre-B lymphomas. Ep-CAM is a Ca[2+] independent homophilic adhesion molecule that is proposed to play roles in the development and normal function of epithelial tissues and in the progression of carcinomas.

Specifications

Specifications

CD326
G8.8
APC
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Ep-CAM; EGP; EGP-2; Egp314; GA733-2; TROP1; Tacsd1; Tacstd1; Ly74; gp40
Affinity Purified
Glycoconjugates from BALB/c mouse-derived TE-71 medullary thymic epithelial cell line
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit