Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD34 Rat anti-Mouse, Clone: MEC14.7, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA182073

Catalog No. MA182073


Add to cart

Description

Description

For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL. Rat anti Mouse CD34 antibody, clone MEC14.7 recognizes the murine CD34 cell surface antigen, which is expressed by endothelial cells and by haematopoietic stem cells.

CD34 is a highly glycosylated, monomeric, 111-115 kDa surface protein that is present on many stem cell populations. CD34 is a stem cell marker although its expression on human hematopoietic stem cells is reversible. CD34 may serve as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation, and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions. CD34 is possibly an adhesion molecule with a putative role for mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Further, CD34 could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. CD34 is thought to have a role in presenting carbohydrate ligands to selectins. The intracellular chain of the CD34 antigen is a site of phosphorylation by activated protein kinase C suggesting a putative role in signal transduction. Diseases associated with CD34 dysfunction include dermatofibrosarcoma and neurofibroma.
Specifications

Specifications

CD34
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q64314
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Rat
IgG2a
100 μg
4°C or -20°C if preferred
Primary
12490
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
MEC14.7
Unconjugated
Cd34
Liquid
CD34
T-end.1, a pMT transformed mouse endothelial cell line
Protein G
RUO
Antibody
Monoclonal
Mouse
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit