Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CD34, PE, Clone: 756510, R&D Systems™
Supplier: R&D Systems FAB72271P
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
CD34 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
947 | |
Human | |
Purified |
Flow Cytometry | |
756510 | |
Flow Cytometry 10 uL/10^6 cells | |
P28906 | |
CD34 | |
Chinese hamster ovary cell line CHO-derived recombinant human CD34, Ser32-Thr290, Accession # P28906 | |
100 Tests | |
Primary | |
Detects human CD34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse, rat, or porcine CD34 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title