Learn More
CD34 Rat anti-Mouse, PE, Clone: MEC 14.7, Invitrogen™ Non-distribution product as customer accommodation.
Rat Monoclonal Antibody
Manufacturer:  Invitrogen RM3604
Catalog No. 50-113-8178
Add to cart
Description
Description
CD34 is a highly glycosylated monomeric 111-115kDa surface protein, which is present on many stem cell populations. It is a well established stem cell marker, though its expression on human hematopoietic stem cells is reversible. CD34 probably serves as a surface receptor that undergoes receptor-mediated endocytosis and regulates adhesion, differentiation and proliferation of hematopoietic stem cells and other progenitors. CD34 expression is likely to represent a specific state of hematopoietic development that may have altered adhering properties with expanding and differentiating capabilities in both in vitro and in vivo conditions.
Specifications
Specifications
CD34
Monoclonal
PE
Cd34
Liquid
Cd34
t-end.1, a polyoma Middle T oncogene (pMT) transformed EC line
Purified
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
MEC 14.7
PBS with BSA and 0.1% sodium azide
Q64314
Mucosialin, RP11-328D5.2, CD34 antigen, hematopoietic progenitor cell antigen CD34
Rat
IgG2a
500μL
4° C
Primary
12490