Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD344 Mouse anti-Human, PE, Clone: CH3A4A7, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 563660

Catalog No. BDB563660


Add to cart

Description

Description

The CH3A4A7 monoclonal antibody reacts with an extracellular epitope on human CD344 which is also known as Frizzled family receptor 4 (FZD4) or Frizzled-4. CD344 is a member of the Frizzled family of proteins within the G protein-coupled receptor (GPCR) superfamily. CD344 is a receptor for Wnt proteins and participates in the activation of the Wnt/beta-catenin pathway (canonical Wnt signaling). Patients with familial exudative vitreoretinopathy (FEVR) carry mutations in Frizzled-4. This disease is characterized by retinal degeneration and hearing loss. On tissues, CD344 is almost ubiquitously expressed. CD344 is highly expressed in adult heart and skeletal muscle.

Specifications

Specifications

CD344
CH3A4A7
FZD4; Frizzled family receptor 4; Frizzled-4; Fz-4; FzE4; hFz4; EVR1; FEVR
Affinity Purified
Human neuroblastoma Cell Line
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit