Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD39 Mouse anti-Human, FITC, Clone: TU66 (also known as Tü 66, Tü66), BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 561444

Catalog No. BDB561444


Add to cart

Description

Description

The TU66 monoclonal antibody reacts with human CD39 also known as ectonucleoside triphosphate diphosphohydrolase 1 (ENTPD1) an ectoenzyme that degrades ATP to AMP. It is a member of the family of ectonucleoside triphosphate dihydrolases (E-NTPDases) known to be involved in regulation of extracellular nucleotide catabolism, controlling the extracellular nucleoside triphosphate pool (NTP). CD39 is expressed on a subset of T cells, B cells and dendritic cells with weak staining of monocytes and granulocytes. Recently, CD39 has been found to be expressed primarly by immune-suppressive Foxp3(+) regulatory T (Treg) cells in both human and mice. In humans, CD39 is restricted to a subset of Foxp3+ regulatory effector/memory-like T cells. In mice, the enzyme is present on most if not all CD4+CD25+ cells and CD39 expression is driven by Foxp3. It is thought that CD39 allows Treg cells to enter inflamed areas where high levels of ATP are present.

Specifications

Specifications

CD39
TU66 (also known as Tü 66, Tü66)
ENTPD1; NTPDase-1; Ecto-ATPase 1; Ecto-ATPDase 1
Affinity Purified
IgG2b, κ
RUO
Primary
Human
Flow Cytometry
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit