Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD39L1/ENTPD2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP144241

Catalog No. NBP144241


Add to cart

Description

Description

CD39L1/ENTPD2 Polyclonal specifically detects CD39L1/ENTPD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

CD39L1/ENTPD2
1.0 mg/mL
Immunohistochemistry, Immunohistochemistry-Paraffin 2.5 ug/ml
Affinity Purified
ENTPD2
Synthetic 18 amino acid peptide from internal region of human CD39L1/ENTPD2.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for ENTPD2. Unless other species reactivities are specified there is no testing data currently available.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.1% Sodium Azide
CD39 antigen-like 1, CD39L1NTPDase 2, CD39-like-1, EC 3.6.1, EC 3.6.1.-, EC 3.6.1.5, Ecto-ATP diphosphohydrolase 2, Ecto-ATPase 2, Ecto-ATPDase 2, ectonucleoside triphosphate diphosphohydrolase 2, NTPDase-2
Rabbit
IgG
0.05 ml
Lipid and Metabolism
Primary
954
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only