Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENTPD3 Rat anti-Mouse, Clone: 568610, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524255

Catalog No. PIMA524255


Add to cart

Description

Description

In Western blots, less than 15% cross-reactivity with Recombinant human (rh) CD39L4 and no cross-reactivity with rhCD39L3, Recombinant mouse (rm) CD39, or rmCD39L2 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

ENTPD3 is similar to E-type nucleotidases (NTPases). NTPases, such as CD39, mediate catabolism of extracellular nucleotides. ENTPD3 contains 4 apyrase-conserved regions which is characteristic of NTPases.
Specifications

Specifications

ENTPD3
Monoclonal
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Lyophilized
ENTPD3
CHO-derived recombit mouse CD39L3/ENTPD3 Gln44-Pro485
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
568610
Unconjugated
Entpd3
CD39L3, HB6, NTPDase-3, CD39 antigen-like 3, CD39-like 3, NTPDase 3, ecto-ATP diphosphohydrolase 3, ecto-ATPDase 3, ecto-ATPase 3, ecto-apyrase 3
Rat
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
215446
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit