Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3e, Polyclonal, Invitrogen™

Catalog No. PIPA130878
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
PIPA130878 1 mL
1 options

Catalog No. PIPA130878

Supplier: Thermo Scientific PA130878

Rabbit Polyclonal Antibody

The protein encoded by this gene is the CD3-epsilon polypeptide, which together with CD3-gamma, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. The epsilon polypeptide plays an essential role in T-cell development. Defects in this gene cause immunodeficiency. This gene has also been linked to a susceptibility to type I diabetes in women.

PA1-30878 detects CD3e from human samples.PA1-30878 has been successfully used in immunohistochemistry (paraffin tissue) applications.The PA1-30878 immunogen is: Synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon chain of human CD3 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3e
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene CD3E
Gene Accession No. P07766
Gene Alias FLJ18683, T3E, TCRE, CD3-epsilon, CD3e antigen, epsilon polypeptide TiT3 complex, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain
Gene Symbols CD3E
Host Species Rabbit
Immunogen Synthetic 13-mer peptide corresponding to aa 156-168 of the epsilon chain of human CD3 protein.
Purification Method Ig fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 916
Target Species Human
Content And Storage Store at 4°C
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.