Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD3e PE-Cy7, Clone: 145-2C11,

Armenian Hamster Monoclonal Antibody

Manufacturer:  MilliporeSigma™ MABF1582

 View more versions of this product

Catalog No. MABF1582MI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifically detects CD3e Clone: 145-2C11 in Mouse samples, and it is validated for Flow Cytometry

The 145-2C11 antibody is specific for mouse CD3e, also known as CD3 epsilon, a 20kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor - CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells. The 145-2C11 antibody is a widely used phenotypic marker for mouse T cells. In addition, binding of 145-2C11 antibody to CD3e can induce cell activation.
Specifications

Specifications

CD3e
145-2C11
Purified Armenian Hamster monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Purified
Armenian Hamster
IgG
100μg
Inflammation, Immunology
Primary
Murine
Flow Cytometry
PE-Cyanine7
P22646
Cd3e
Proprietary immunogen
Purified by size-exclusion chromatography
RUO
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only