Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD3e PerCP-Cy5.5, Clone: 145-2C11,

Armenian Hamster Monoclonal Antibody

$153.50

Specifications

Antigen CD3e
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation Purified Armenian Hamster monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF1583MI
View Documents
MilliporeSigma™
MABF1583
25μg
Each for $153.50
Add to cart
 
MABF1584MI
View Documents
MilliporeSigma™
MABF1584
100μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifically detects CD3e Clone: 145-2C11 in Mouse samples, and it is validated for Flow Cytometry

The 145-2C11 antibody is specific for mouse CD3e, also known as CD3 epsilon, a 20kDa subunit of the T cell receptor complex, along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor - CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. Such interactions promote T cell receptor signaling (T cell activation) and can result in a number of cellular responses including proliferation, differentiation, production of cytokines or activation-induced cell death. CD3 is differentially expressed during thymocyte-to-T cell development and on all mature T cells.The 145-2C11 antibody is a widely used phenotypic marker for mouse T cells. In addition, binding of 145-2C11 antibody to CD3e can induce cell activation. A recent publication of the crystal structure of a murine CD3e-mitogenic antibody complex provides further insight into the action of commonly used agonist antibodies (Fernandes, R.A. et al. 2012. J. Biol. Chem. 287: 13324-13335).
Specifications

Specifications

CD3e
Monoclonal
Purified Armenian Hamster monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Flow Cytometry
IgG
P22646
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Inflammation, Immunology
Primary
Purified
145-2C11
Armenian Hamster
Proprietary immunogen
Cd3e
RUO
Purified by size-exclusion chromatography
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit