Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD4 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
$138.04 - $242.44
Specifications
Antigen | CD4 |
---|---|
Clone | GK1.5 |
Dilution | Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 5 ug/mL |
Applications | Flow Cytometry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB554G025
|
R&D Systems
FAB554G025 |
25 Tests |
Each for $138.04
|
|
|||||
FAB554G100
|
R&D Systems
FAB554G100 |
100 Tests |
Each for $242.44
|
|
|||||
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD4 | |
Flow Cytometry 5 uL/10^6 cells, Immunocytochemistry 5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
920 | |
Mouse CTL clone V4 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
GK1.5 | |
Flow Cytometry, Immunocytochemistry | |
Alexa Fluor 488 | |
Rat | |
Mouse | |
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
CD4 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Reacts with mouse CD4, an antigen co-receptor on the Tácell surface which interacts with MHCáII molecules on antigen presenting cells. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title