Learn More
Mouse CD4 Alexa Fluor 647-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB554R025
 View more versions of this product
Catalog No. FAB554R025
Add to Cart
This item is not returnable. View return policy
Description
Description
CD4 Monoclonal specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD4
Monoclonal
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD4
Mouse CTL clone V4
25 Tests
Primary
Reacts with mouse CD4, an antigen co-receptor on the Tácell surface which interacts with MHCáII molecules on antigen presenting cells.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2b
Flow Cytometry
GK1.5
Flow Cytometry 5 uL/10^6 cells
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
920
Mouse
Supernatant
Product Suggestions
Product Suggestions