anti-CD4-Biotin; Clone: 13B8.2; Beckman Coulter™

$413.11

Specifications

Antigen CD4-Biotin
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Host Species Mouse
Clone 13B8.2
View More Specs
  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability    

COIM0704

Beckman Coulter
IM0704
0.2mg Biotin Each for $413.11
Check Availability
Add to Cart
Descriptions & Specifications

Related Products