Learn More
CD4/CD25, FITC, PE, Clone: 7E14, ITYV, Antibody Cocktail, Invitrogen™
Antibody Cocktail
Manufacturer:  Invitrogen MA112116
Catalog No. 01-670-835
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The CD4 antigen is involved in the recognition of the type II MHC antigen. It is also a receptor for HIV. It is present on most T helper cells and normal thymocytes. The cytoplasmic portion of CD4 is associated with p56lck tyrosine kinase. CD4 expression is commonly found in human lymph nodes and tonsils. Natural killer (NK) cells in teleosts and their evolutionary homologue are a subpopulation of lymphocytes with properties that distinguish them from either B- or T-cells. NK cells are important effectors of innate immunity where they release cytokines, which in turn up-regulate other immunological functions. Monoclonal antibodies have been used to identify different surface antigens present on NK cells. These surface antigens have not only been used to identify NK cells, but also their functionally distinct subsets. The Cluster of Differentiation (CD) nomenclature was established to standardize the naming of NK cells. To date, a total of 151 CD clusters and subclusters have been identified.
This antibody is a monoclonal mixture of FITC-conjugated CD4 (clone 7E14, isotype IgG1) and PE-conjugated CD25 (clone ITYV, isotype IgG1).
Specifications
Specifications
CD4/CD25
Cocktail
FITC, PE
CD4
Liquid
CD4, IL2RA
human peripheral blood T lymphocytes and PHA-activated human T-lymphocytes
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
7E14, ITYV, ITYV
PBS with 0.2% carrier protein and 0.08% sodium azide
P01589, P01730
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4, IL-2R alpha, IL2 RA antibody, Il2ra antibody, Interleukin 2 receptor alpha chain antibody, TAC antigen antibody, IL-2R alpha chain
Murine
IgG1
100 Tests
4° C, do not freeze, do not freeze
Primary
3559, 920