Learn More
CD4 Rat anti-Mouse, Unlabeled, Clone: GK1.5, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 553727
Description
The GK1.5 clone has been reported to react with the CD4 (L3T4) differentiation antigen expressed on most thymocytes, a subpopulation of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells), and a subset of NK-T cells. In addition, CD4 has also been reported to be detectable on pluripotent hematopoietic stem cells, bone marrow myeliod and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 has also been reported to be expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. GK1.5 mAb reportedly blocks binding of the RM4-5 (Cat. No. 553046/553047) and H129.19 (Cat. No. 553650/553651), but not RM4-4 (Cat. No. 553055) antibodies. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD4 | |
Monoclonal | |
0.5mg/mL | |
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
GK1.5 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse CTL clone V4 | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2b κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.