Learn More
anti-CD4, Clone: GK1.5, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB554V025
 View more versions of this product
Catalog No. FAB554V025
Add to cart
Specifications
Specifications
CD4
GK1.5
Flow Cytometry 5 uL/10^6 cells
Supernatant
CD4
Mouse CTL clone V4
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
920
Mouse
Flow Cytometry
Alexa Fluor 405
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Rat
IgG2b
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Reacts with mouse CD4, an antigen co-receptor on the Tácell surface which interacts with MHCáII molecules on antigen presenting cells.