Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD4/HLA-DR Antibody Cocktail, FITC, PE, Invitrogen™
Antibody Cocktail
Supplier: Thermo Scientific MA112281
Description
This antibody is a monoclonal mixture of FITC-conjugated CD4 (clone 7E14, isotype IgG1) and PE-conjugated HLADR (clone 423L, isotype IgG2a).
CD4/HLA-DR Cocktail antibody specifically detects CD4/HLA-DR in Human samples. It is validated for Flow Cytometry.Specifications
CD4/HLA-DR | |
Cocktail | |
FITC/PE | |
CD4 | |
CD4; CD4 antigen (p55); CD4 antigen p55; CD4 molecule; CD4 receptor; CD4mut; CD74; CD74 antigen; CD74 antigen (invariant polypeptide of major histocompatibility complex, class II antigen-associated); CD74 molecule; CD74 molecule, major histocompatibility complex, class II invariant chain; DHLAG; gamma chain of class II antigens; HLA class II histocompatibility antigen gamma chain; HLADG; HLA-DR antigens-associated invariant chain; HLA-DR-gamma; Ia antigen-associated invariant chain; Ia-associated invariant chain; Ia-GAMMA; II; MHC HLA-DR gamma chain; p33; T-cell surface antigen T4/Leu-3; T-cell surface glycoprotein CD4 | |
Mouse | |
Protein A/G | |
RUO | |
920 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
7E14, 423L, 423L | |
PBS with 0.2% carrier protein and 0.08% sodium azide | |
P01730 | |
CD4 | |
human peripheral blood T lymphocytes | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction