Learn More
anti-CD4, PE, Clone: 11830, R&D Systems™
Mouse Monoclonal Antibody has been used in 6 publications
Manufacturer:  R&D Systems FAB3791P100
 View more versions of this product
Catalog No. FAB3791P100
Add to cart
This item is not returnable. View return policy
Description
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD4
11830
Flow Cytometry 10 uL/10^6 cells
P01730
CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
IgG2a
100 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human CD4 in direct ELISAs.
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Purified
CD4
Recombinant Human CD4. Extracellular Domain Accession # P01730
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
920
Human