Learn More
CD4 Mouse anti-Human, PE, Clone: MDX.2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112771
Catalog No. 01-674-054
Add to cart
Description
Description
The CD4 antigen is involved in the recognition of MHC class II molecules and is a co-receptor for HIV. CD4 is primarily expressed in a subset of T-lymphocytes, also referred to as T helper cells, but may also be expressed by other cells in the immune system, such as monocytes, macrophages, and dendritic cells. At the tissue level, CD4 expression may be detected in thymus, lymph nodes, tonsils, and spleen, and also in specific regions of the brain, gut, and other non-lymphoid tissues. CD4 functions to initiate or augment the early phase of T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase, Lck. It may also function as an important mediator of direct neuronal damage in infectious and immune-mediated diseases of the central nervous system. Multiple alternatively spliced transcripts have been identified in this gene [RefSeq, July 2017].
Specifications
Specifications
CD4
Monoclonal
PE
CD4
Liquid
CD4
human peripheral blood T lymphocytes
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MDX.2
PBS with 0.2% carrier protein and 0.08% sodium azide
P01730
CD4 antigen p55, CD4 receptor, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4
Mouse
IgG1
100 Tests
4° C
Primary
920