Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD4 Red Fluor 710, Clone: RM4-5,
Rat Monoclonal Antibody
$184.00 - $328.00
Specifications
Antigen | CD4 |
---|---|
Clone | RM4-5 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Red Fluor 710 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1577MI
|
MilliporeSigma™
MABF1577 |
25 μg |
Each for $184.00
|
|
|||||
MABF1578MI
|
MilliporeSigma™
MABF1578 |
100 μg |
Each for $328.00
|
|
|||||
Description
Specifically detects CD4 Clone: RM4-5 in Mouse samples, and it is validated for Flow Cytometry
The RM4-5 antibody reacts with mouse CD4, a 55kDa protein which acts as a co-receptor for the T cell receptor (TCR) in its interaction with MHC Class II molecules on antigen-presenting cells. The extracellular domain of CD4 binds to the beta2-domain of MHC Class II, while its cytoplasmic tail provides a binding site for the tyrosine kinase lck, facilitating the signaling cascade that initiates T cell activation. CD4 is typically expressed on thymocytes, certain mature T cell populations such as Th17 and T regulatory (Treg) cells, as well as on dendritic cells.The RM4-5 antibody is widely used as a phenotypic marker for CD4 expression. If used together, the RM4-5 antibody and an alternative antibody, Anti-Mouse CD4 clone GK1.5, will 'compete' for binding, i.e. RM4-5 antibody is able to block GK1.5 antibody binding to cells. In contrast, RM4-5 antibody does not block the binding of Anti-Mouse CD4 clone RM4-4 to cells.Specifications
CD4 | |
Flow Cytometry | |
Red Fluor 710 | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Cd4 | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
RM4-5 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P06332 | |
Proprietary immunogen | |
Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title