Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Mouse anti-Human, Unlabeled, Clone: 41, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611362

Catalog No. BDB611362


Add to cart

Description

Description

CD40, a member of the TNF receptor family, contains a cysteine-rich N-terminal domain and a Ser/Thr-rich region preceding the transmembrane domain. On B cells, signaling through CD40 induces cell growth and differentiation, mediates cell survival within the germinal center, and upregulates the expression of costimulatory and adhesion molecules, such as B7.1, B7.2, and ICAM-1. The interaction of CD40 on B cells and CD40L on activated CD4+ T cells is essential for immune functions, such as immuoglobulin class switching. Signal transduction through CD40 pathways involves interaction with proteins such as TRAFs (TRAF2, TRAF3, TRAF5, and TRAF6); Jak 3; and Tyr phosphorylation of proteins, such as Lyn, Syk, PI-3-kinase, STAT3, and STAT5. In TRAF2-deficient mice, CD40-mediated B cell proliferation and NFκB activation are defective. Ku70 and Ku80 associate with the membrane-proximal region of CD40 in human primary B cells and the engagement of CD40 leads translocation of Ku proteins to the nucleus. Thus, CD40 interacts with a variety of signal transducers which mediate its role in B cell survival, growth, differentiation, and immunoglobulin class switching.

Specifications

Specifications

CD40
41
Unconjugated
Affinity Purified
Human CD40 aa. 71-187
50μg
Immunology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit