Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD40 Goat anti-Human, Mouse, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA124879

Catalog No. PIPA124879


Add to cart

Description

Description

PA1-24879 detects CD40 from mouse samples. PA1-24879 has been successfully used in ELISA and Western blot procedures. The PA1-24879 immunogen is recombit, extracellular domain of mouse CD40, expressed in mouse NSO cells.

The CD40 antigen is a single chain glycoprotein that is known to be a member of the tumor necrosis factor/nerve growth factor superfamily and shows a significant homology to the Hodgkin ft.s disease associated antigen, CD30. CD40 is present on all B cells except plasma cells and is also found on some epithelial cells, carcinomas and lymphoid dendritic cells. CD40 has been found to be essential in mediating a broad variety of immune and inflammatory responses including T cell-dependent immunoglobulin class switching, memory B cell development, and germinal center formation. AT-hook transcription factor AKNA is reported to coordinately regulate the expression of CD40, which may be important for homotypic cell interactions. Adaptor protein TNFR2 interacts with CD40 and serves as a mediator of the signal transduction. The interaction of CD40 and its ligand is found to be necessary for amyloid-beta-induced microglial activation, and thus is thought to be an early event in Alzheimer disease pathogenesis. Two alternatively spliced transcript variants of CD40 encoding distinct isoforms have been reported. Diseases associated with CD40 dysfunction include Type 3 Hyper-Igm immunodeficiency and CD40 ligand deficiency.
Specifications

Specifications

CD40
Polyclonal
Unconjugated
CD40
Liquid
CD40
Recombit, extracellular domain of mouse CD40, expressed in mouse NSO cells.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Western Blot
0.1 mg/mL
PBS with 1% BSA and no preservative; pH 7.4
P25942, P27512
Bp50, CDW40, MGC9013, TNFRSF5, p50
Goat
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
21939, 958
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit