Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD154 (CD40 Ligand), PE, clone: 40804, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523528

Catalog No. PIMA523528

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.

In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CD27 Ligand, rhCD30 Ligand, or recombinant mouse CD40 Ligand is observed.
Specifications

Specifications

CD154 (CD40 Ligand)
Monoclonal
PE
CD40LG
Liquid
CD40LG
E. coli-derived recombinant human CD40 Ligand/TNFSF5 Leu50-Leu261
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
40804
Buffered saline solution with BSA and 0.09% sodium azide
P29965
RP23-153G22.3, CD154, CD40-L, Cd40l, HIGM1, IGM, IMD3, Ly-62, Ly62, T-BAM, TRAP, Tnfsf5, gp39, T-cell antigen Gp39, TNF-related activation protein, tumor necrosis factor ligand superfamily member 5
Murine
IgG2B
100 Tests
Store at 4°C, store in the dark, do not freeze
Primary
959
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit