Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD41 Rat anti-Mouse, BUV395, Clone: MWReg30, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564056

Catalog No. BDB564056


Add to cart

Description

Description

The MWReg30 antibody reacts with glycoprotein (gp) IIb, also called integrin α IIb chain (CD41), that associates with integrin β 3 chain (gpIIIa or CD61) to form the gpIIb/IIIa (CD41/CD61) complex. CD41/CD61 is expressed on platelets, megakaryocytes, and early hematopoietic progenitors. The integrin complex binds to fibrinogen, fibronectin, vitronectin, von Willebrand factor, and thrombospondin. It is important for platelet adhesion and aggregation, and it may play a role in osteolytic tumor metastasis.

Specifications

Specifications

CD41
MWReg30
Brilliant Ultraviolet 395
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Rat
IgG1, κ
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Integrin α IIb chain
Affinity Purified
Mouse Platelets
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit