Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD42a, Biotin, clone: ESS, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112215

Catalog No. 01-670-923

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.

Specifications

Specifications

CD42a/GP9
Monoclonal
Biotin
GP9
Purified
GP9
White blood cells.
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
ESS
PBS with 0.2% carrier protein and 0.08% sodium azide
P14770
CD42a, GPIX, GP-IX, glycoprotein 9, platelet glycoprotein IX, Platelet GPIX
Murine
IgG1
100 Tests
Store at 4°C
Primary
2815
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit