Learn More
Thermo Scientific Lab Vision CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.
Manufacturer: Thermo Scientific MS679P
Description
Obtain accurate, reproducible results in immunohistochemistry and flow cytometry experiments with Thermo Scientific CD43/T-Cell Marker Ab-3, Mouse Monoclonal Antibody.
Host Species: Mouse
Clone: 84-3C1
Isotype: IgG1
Species Reactivity: Human. Does not react with rat. Others not tested.
Epitope: Not determined
Immunogen: Stimulated human leukocytes
Molecular Weight: 95/115/135 kDa (depending upon extent of glycosylation)
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD43/T-Cell Marker Ab-3 | |
Monoclonal | |
200μg/mL | |
Purified Ab with BSA and Azide | |
Murine | |
IgG1 | |
RUO | |
Primary | |
Monoclonal |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
84-3C1 | |
Unconjugated | |
Concentrated | |
Stimulated human leukocytes | |
1mL | |
T-Cell | |
Primary | |
Human |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.