Learn More
anti-CD45.1, Alexa Fluor 700, Clone: A20, R&D Systems™
Manufacturer:  R&D Systems FAB8374N025
 View more versions of this product
Catalog No. FAB8374N025
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD45.1
A20
Flow Cytometry 5 uL/10^6 cells
Purified
PTPRC
SJL mouse thymocytes and splenocytes
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5788
Mouse
Flow Cytometry
Alexa Fluor 700
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
B220, CD45, CD45 antigen, CD45.1, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, Ly-5.1, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
IgG2a
25 Tests
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse cells expressing the CD45.1 allotype in flow cytometry. Clone A20 does not detect the mouse CD45.2 alloantigen.