Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD45.1; Clone: A20; BD

Manufacturer:  BD Biosciences 563754

Catalog No. BDB563754


Add to cart

Description

Description

Host Species: Mouse
Clone: A20
Isotype: IgG2a, κ
Species Reactivity: Ms
Immunogen: SJL Mouse thymocytes and splenocytes

Flow cytometry

Specifications

Specifications

CD45.1
0.2mg/mL
Murine
50μg
Primary
Murine
A20
Affinity Purified
SJL Mouse thymocytes and splenocytes
RUO
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit